Assay Details
Gene Symbol: Atp2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134462.1 | NP_604457.1 | 11 - 12 | 1090 | 63 | |
XM_008772595.1 | XP_008770817.1 | 5 - 6 | 3070 | 63 | ||
XM_008772596.1 | XP_008770818.1 | 5 - 6 | 1032 | 63 | ||
GenBank mRNA | AF484685.1 | - | 11 - 12 | 1090 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
