Assay Details
| Gene Symbol: Zfp394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145724.1 | NP_663776.1 | 2 - 3 | 690 | 83 | |
| GenBank mRNA | AF052042.1 | - | 2 - 3 | 690 | 83 | |
| BC100140.1 | - | 2 - 3 | 684 | 7442414 | 83 | |
More Information
Set Membership: |
|

Zfp394
Genomic Map