Assay Details
Gene Symbol: Soat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153728.2 | NP_714950.2 | 6 - 7 | 992 | 64 | |
GenBank mRNA | AB075946.1 | - | 6 - 7 | 724 | 64 | |
AB101480.1 | - | 6 - 7 | 992 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
