Assay Details
Gene Symbol: Odf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007670.1 | NP_001007671.1 | 3 - 4 | 592 | 94 | |
XM_006246698.2 | XP_006246760.1 | 3 - 4 | 1014 | 94 | ||
GenBank mRNA | BC079319.1 | - | 3 - 4 | 592 | 7135137 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
