Assay Details
| Gene Symbol: Ubr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007705.1 | NP_001007706.1 | 1 - 2 | 178 | 67 | |
| XM_006240475.2 | XP_006240537.1 | 1 - 2 | 234 | 67 | ||
| XM_006240476.2 | XP_006240538.1 | 1 - 2 | 234 | 67 | ||
| GenBank mRNA | BC081966.1 | - | 1 - 2 | 178 | 7125402 | 67 |
More Information
Set Membership: |
|

Ubr7
Genomic Map