Assay Details
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007720.1 | NP_001007721.1 | 1 - 2 | 86 | 94 | |
GenBank mRNA | AF110267.1 | - | 1 - 2 | 94 | 94 | |
BC078731.1 | - | 1 - 2 | 86 | 7112220 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
