Assay Details
Gene Symbol: Zfp472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182823.1 | NP_877975.1 | 2 - 3 | 226 | 108 | |
GenBank mRNA | AY195874.1 | - | 2 - 3 | 226 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
