Assay Details
Gene Symbol: Art2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198735.2 | NP_942030.1 | 1 - 2 | 87 | 103 | |
XM_008759696.1 | XP_008757918.1 | 1 - 2 | 936 | 103 | ||
XM_008759697.1 | XP_008757919.1 | 2 - 3 | 1304 | 103 | ||
XM_008759698.1 | XP_008757920.1 | 4 - 5 | 676 | 103 | ||
XM_008759702.1 | XP_008757924.1 | 1 - 2 | 936 | 103 | ||
GenBank mRNA | BC099070.1 | - | 1 - 2 | 87 | 7366134 | 103 |
FQ230306.1 | - | 3 - 4 | 423 | 103 | ||
M85193.1 | - | 1 - 2 | 64 | 103 | ||
X52082.1 | - | 1 - 2 | 57 | 103 | ||
X99122.1 | - | 4 - 5 | 681 | 103 | ||
X99123.1 | - | 2 - 3 | 137 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
