Assay Details
Gene Symbol: Syngr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025644.1 | NP_001020815.1 | 4 - 5 | 690 | 71 | |
XM_006229034.2 | XP_006229096.1 | 4 - 5 | 906 | 71 | ||
GenBank mRNA | BC098938.1 | - | 4 - 5 | 690 | 7455020 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
