Assay Details
Gene Symbol: Ppp1r11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212542.3 | NP_997707.2 | 2 - 3 | 377 | 71 | |
XM_006255870.1 | XP_006255932.1 | 2 - 3 | 442 | 71 | ||
GenBank mRNA | BC079252.1 | - | 2 - 3 | 346 | 7133103 | 71 |
EX488885.1 | - | 2 - 3 | 371 | 71 | ||
FM088354.1 | - | 2 - 3 | 376 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
