Assay Details
| Gene Symbol: Aimp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037348.1 | NP_001032425.1 | 2 - 3 | 373 | 91 | |
| XM_006248863.2 | XP_006248925.1 | 2 - 3 | 320 | 91 | ||
| GenBank mRNA | BC107947.1 | - | 2 - 3 | 373 | 7386574 | 91 |
More Information
Set Membership: |
|

Aimp2
Genomic Map