Assay Details
| Gene Symbol: Defb52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037524.1 | NP_001032613.1 | 2 - 3 | 117 | 80 | |
| XM_006253368.2 | XP_006253430.1 | 2 - 3 | 173 | 80 | ||
| GenBank mRNA | DQ012091.1 | - | 2 - 3 | 117 | 80 | |
More Information
Set Membership: |
|

Defb52
Genomic Map