Assay Details
| Gene Symbol: Cdc42se1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039044.1 | NP_001034133.1 | 2 - 3 | 330 | 57 | |
| GenBank mRNA | BC091418.1 | - | 2 - 3 | 330 | 7365287 | 57 |
More Information
Set Membership: |
|

Cdc42se1
Genomic Map