Assay Details
Gene Symbol: Prl5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044702.1 | NP_001038167.1 | 3 - 4 | 375 | 99 | |
GenBank mRNA | DQ329280.1 | - | 3 - 4 | 375 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
