Assay Details
| Gene Symbol: B3galt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105887.1 | NP_001099357.1 | 2 - 3 | 590 | 76 | |
| XM_008768569.1 | XP_008766791.1 | 2 - 3 | 2645 | 76 | ||
| XM_008768570.1 | XP_008766792.1 | 2 - 3 | 742 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105887.1, XM_008768569.1, XM_008768570.1 transcripts.
Set Membership: |
|

B3galt5
Genomic Map