Assay Details
| Gene Symbol: Prl7a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008341.1 | NP_001008342.1 | 2 - 3 | 265 | 122 | |
| GenBank mRNA | AY741310.1 | - | 2 - 3 | 265 | 122 | |
More Information
Set Membership: |
|

Prl7a4
Genomic Map