Assay Details
| Gene Symbol: C1qa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008515.1 | NP_001008515.1 | 1 - 2 | 29 | 55 | |
| GenBank mRNA | BC086605.1 | - | 1 - 2 | 29 | 7316796 | 55 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC086605.1, NM_001008515.1 transcripts.
Set Membership: |
|

C1qa
Genomic Map