Assay Details
| Gene Symbol: RGD1310553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008517.1 | NP_001008517.1 | 1 - 2 | 113 | 105 | |
| GenBank mRNA | BC086514.1 | - | 1 - 2 | 113 | 7134744 | 105 |
More Information
Set Membership: |
|

RGD1310553
Genomic Map