Assay Details
| Gene Symbol: RGD1310553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008517.1 | NP_001008517.1 | 2 - 3 | 282 | 93 | |
| GenBank mRNA | BC086514.1 | - | 2 - 3 | 282 | 7134744 | 93 |
More Information
Set Membership: |
|

RGD1310553
Genomic Map