Assay Details
Gene Symbol: Zfp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106691.1 | NP_001100161.1 | 1 | 415 | 87 | |
XM_006239150.2 | XP_006239212.1 | 4 | 1406 | 87 | ||
XM_006239152.2 | XP_006239214.1 | 3 | 1351 | 87 | ||
XM_008764217.1 | XP_008762439.1 | 3 | 766 | 87 | ||
XM_008764218.1 | XP_008762440.1 | 4 | 674 | 87 | ||
XM_008764219.1 | XP_008762441.1 | 3 | 882 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
