Assay Details
| Gene Symbol: Magee2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106941.1 | NP_001100411.1 | 1 | 1614 | 80 | |
| XM_006257144.2 | XP_006257206.1 | 3 | 2967 | 80 | ||
| XM_008773341.1 | XP_008771563.1 | 2 | 3154 | 80 | ||
| XM_008773342.1 | XP_008771564.1 | 3 | 3027 | 80 | ||
More Information
Set Membership: |
|

Magee2
Genomic Map