Assay Details
| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107757.1 | NP_001101227.1 | 2 - 3 | 166 | 71 | |
| XM_006234707.2 | XP_006234769.1 | 3 - 4 | 540 | 71 | ||
| XM_008762093.1 | XP_008760315.1 | 3 - 4 | 540 | 71 | ||
More Information
Set Membership: |
|

Aven
Genomic Map