Assay Details
| Gene Symbol: Nat14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108466.1 | NP_001101936.1 | 1 - 2 | 436 | 85 | |
| XM_006228268.2 | XP_006228330.1 | 2 - 3 | 386 | 85 | ||
| XM_006228269.2 | XP_006228331.1 | 2 - 3 | 383 | 85 | ||
| XM_006228270.1 | XP_006228332.1 | 2 - 3 | 337 | 85 | ||
| XM_006228271.1 | XP_006228333.1 | 2 - 3 | 334 | 85 | ||
| XM_006228272.2 | XP_006228334.1 | 2 - 3 | 690 | 85 | ||
| XM_006228273.2 | XP_006228335.1 | 2 - 3 | 687 | 85 | ||
More Information
Set Membership: |
|

Nat14
Genomic Map