Assay Details
| Gene Symbol: Rprml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109481.1 | NP_001102951.1 | 1 | 221 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109481.1 transcripts.
Set Membership: |
|

Rprml
Genomic Map