Assay Details
| Gene Symbol: LOC688765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109512.1 | NP_001102982.1 | 1 | 924 | 66 | |
| XM_006253209.2 | XP_006253271.1 | 1 | 1267 | 66 | ||
More Information
Set Membership: |
|

LOC688765
Genomic Map