Assay Details
| Gene Symbol: Chchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013431.1 | NP_001013449.1 | 3 | 924 | 80 | |
| GenBank mRNA | BC091407.1 | - | 3 | 924 | 7324948 | 80 |
| FQ220478.1 | - | 3 | 789 | 80 | ||
| FQ220795.1 | - | 3 | 819 | 80 | ||
| FQ229282.1 | - | 3 | 789 | 80 | ||
More Information
Set Membership: |
|

Chchd4
Genomic Map