Assay Details
| Gene Symbol: Hbg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172093.1 | NP_742090.1 | 3 | 326 | 120 | |
| GenBank mRNA | BC157810.1 | - | 3 | 360 | 9027502 | 120 |
| FQ221361.1 | - | 3 | 378 | 120 | ||
| FQ222999.1 | - | 3 | 373 | 120 | ||
| FQ228385.1 | - | 3 | 380 | 120 | ||
| FQ228657.1 | - | 3 | 376 | 120 | ||
More Information
Set Membership: |
|

Hbg1
Genomic Map