Assay Details
Gene Symbol: RGD1560010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107685.1 | NP_001101155.1 | 2 | 1418 | 111 | |
GenBank mRNA | BC162053.1 | - | 2 | 1382 | 7113318 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
