Assay Details
| Gene Symbol: Ppp1r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138521.1 | NP_612530.1 | 5 - 6 | 869 | 60 | |
| XM_006247344.2 | XP_006247406.1 | 2 - 3 | 1367 | 60 | ||
| GenBank mRNA | AF281661.1 | - | 1 - 2 | 194 | 60 | |
| AY601872.1 | - | 5 - 6 | 869 | 60 | ||
| BC078954.1 | - | 5 - 6 | 799 | 7110316 | 60 | |
More Information
Set Membership: |
|

Ppp1r1b
Genomic Map