Assay Details
| Gene Symbol: Thtpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251970.2 | XP_006252032.1 | 1 - 2 | 2878 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Thtpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251970.2 | XP_006252032.1 | 1 - 2 | 2878 | 83 | |
Set Membership: |
|