Assay Details
| Gene Symbol: Pcdhga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772071.1 | XP_008770293.1 | 1 | 2697 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhga4| Gene Symbol: Pcdhga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772071.1 | XP_008770293.1 | 1 | 2697 | 67 | |
Set Membership: |
|