Assay Details
| Gene Symbol: Fam212b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233088.2 | XP_006233150.1 | 1 - 2 | 213 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam212b| Gene Symbol: Fam212b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233088.2 | XP_006233150.1 | 1 - 2 | 213 | 58 | |
Set Membership: |
|