Assay Details
Gene Symbol: Fancd2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237051.2 | XP_006237113.1 | 2 - 3 | 785 | 76 | |
XM_006237052.2 | XP_006237114.1 | 2 - 3 | 1386 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
