Assay Details
| Gene Symbol: Fam187b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001069328.3 | XP_001069328.2 | 1 - 2 | 1001 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam187b| Gene Symbol: Fam187b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001069328.3 | XP_001069328.2 | 1 - 2 | 1001 | 60 | |
Set Membership: |
|