Assay Details
| Gene Symbol: LOC684828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771681.1 | XP_008769903.1 | 1 | 621 | 98 | |
| Gene Symbol: LOC100910366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771686.1 | XP_008769908.1 | 2 | 637 | 98 | |
More Information
Set Membership: |
|

LOC684828
Genomic Map