Assay Details
Gene Symbol: Lor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761253.1 | XP_008759475.1 | 2 | 2671 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761253.1 | XP_008759475.1 | 2 | 2671 | 60 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |