Assay Details
Gene Symbol: LOC499235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_574528.5 | XP_574528.1 | 1 | 100 | 129 | |
GenBank mRNA | AY539892.1 | - | 1 | 100 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
