Assay Details
| Gene Symbol: Khdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766940.1 | XP_008765162.1 | 1 | 319 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008766940.1 transcripts.
Set Membership: |
|

Khdc1b
Genomic Map