Assay Details
Gene Symbol: LOC100910057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244225.2 | XP_006244287.1 | 7 | 997 | 127 | |
XM_006244227.2 | XP_006244289.1 | 8 | 1034 | 127 | ||
XM_006244228.2 | XP_006244290.1 | 8 | 1339 | 127 | ||
XM_006244229.2 | XP_006244291.1 | 8 | 1029 | 127 | ||
XM_006244230.2 | XP_006244292.1 | 8 | 1334 | 127 | ||
XM_006244231.2 | XP_006244293.1 | 7 | 975 | 127 | ||
XM_006244232.2 | XP_006244294.1 | 7 | 955 | 127 | ||
XM_006244233.2 | XP_006244295.1 | 7 | 1260 | 127 | ||
XM_006244236.2 | XP_006244298.1 | 5 | 693 | 127 | ||
XM_008766781.1 | XP_008765003.1 | 8 | 1034 | 127 | ||
XM_008766782.1 | XP_008765004.1 | 8 | 1017 | 127 | ||
XM_008766783.1 | XP_008765005.1 | 8 | 1012 | 127 | ||
XM_008766784.1 | XP_008765006.1 | 8 | 990 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
