Assay Details
Gene Symbol: Ddit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U36994.1 | - | 1 | 46 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U36994.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
