Assay Details
| Gene Symbol: MAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001180313.1 | NP_010288.1 | 1 | 1052 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAF1| Gene Symbol: MAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001180313.1 | NP_010288.1 | 1 | 1052 | 101 | |
Set Membership: |
|