Assay Details
| Gene Symbol: MSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001180513.3 | NP_010491.4 | 1 | 2052 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MSC2| Gene Symbol: MSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001180513.3 | NP_010491.4 | 1 | 2052 | 102 | |
Set Membership: |
|