Assay Details
| Gene Symbol: ESC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001180671.1 | NP_010650.1 | 1 | 1048 | 101 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESC2| Gene Symbol: ESC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001180671.1 | NP_010650.1 | 1 | 1048 | 101 | |
Set Membership: | 
		   
					
						
							 |