Assay Details
| Gene Symbol: RPN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001180735.3 | NP_010715.3 | 1 | 747 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPN9| Gene Symbol: RPN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001180735.3 | NP_010715.3 | 1 | 747 | 105 | |
Set Membership: |
|