Assay Details
Gene Symbol: SPR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001179005.1 | NP_011040.1 | 1 | 450 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
