Assay Details
| Gene Symbol: HCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182079.1 | NP_013293.1 | 1 | 655 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCR1| Gene Symbol: HCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182079.1 | NP_013293.1 | 1 | 655 | 102 | |
Set Membership: |
|