Assay Details
Gene Symbol: ERF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182133.1 | NP_013347.1 | 1 | 951 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
