Assay Details
| Gene Symbol: CYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001183484.1 | NP_014708.1 | 1 | 849 | 82 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  Detect Genomic DNA  Saccharomyces cerevisiae  Both primers and probe map within a single exon | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 CYT1
CYT1 Genomic Map
Genomic Map Back To Top
 Back To Top