Assay Details
| Gene Symbol: RPA190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001183761.3 | NP_014986.3 | 1 | 4853 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPA190| Gene Symbol: RPA190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001183761.3 | NP_014986.3 | 1 | 4853 | 103 | |
Set Membership: |
|