Assay Details
| Gene Symbol: SEC22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070245.1 | XP_020925904.1 | 561 | 75 | ||
| XM_021070246.1 | XP_020925905.1 | 802 | 75 | |||
| XM_021070247.1 | XP_020925906.1 | 947 | 75 | |||
| XM_021070249.1 | XP_020925908.1 | 455 | 75 | |||
| XM_021070250.1 | XP_020925909.1 | 455 | 75 | |||
| XM_021070251.1 | XP_020925910.1 | 455 | 75 | |||
More Information
Set Membership: |
|

SEC22A
Genomic Map